MilliporeSigma
  • The fate of Microcystis aeruginosa cells during the ferric chloride coagulation and flocs storage processes.
The fate of Microcystis aeruginosa cells during the ferric chloride coagulation and flocs storage processes.
Environmental technology (2014-09-23)
Xiuqing Li, Haiyan Pei, Wenrong Hu, Panpan Meng, Feng Sun, Guixia Ma, Xiangchao Xu, Yuezhong Li
ABSTRACT
Microcystis blooms could cause severe problems for drinking water supplies with their associated microcystins (MCs). As the majority of MCs are retained inside the cells, the effective removal of the intact Microcystis cells to avoid the release of additional MCs plays an important role in drinking water treatment. This study evaluated the effect of ferric chloride (FeCl3) coagulation and the flocs storage process on the integrity of Microcystis aeruginosa cells and the intracellular MCs release (and possible degradation) in both processes. Multiple analysis techniques including scanning electron microscopy and chlorophyll fluorescence were used to assess the integrity of M. aeruginosa. In the coagulation process, the coagulant dosage and mechanical actions caused no cell damage, and all the cells remained intact. Furthermore, 100 mg/L FeCl3 was effective in removing the extracellular MCs. In the flocs storage process, a number of intracellular MCs were released into the supernatant, but the cells remained viable up to 10 d.
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Millipore
TDA Reagent, suitable for microbiology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00