Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid, 99.999%
Sign Into View Organizational & Contract Pricing USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Orcinol, 97%
Sign Into View Organizational & Contract Pricing Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sign Into View Organizational & Contract Pricing