Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
Sign Into View Organizational & Contract Pricing Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur™, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Carbon, mesoporous
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sign Into View Organizational & Contract Pricing Supelco
N,O-Bis(trimethylsilyl)acetamide, for GC derivatization, LiChropur™, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Sign Into View Organizational & Contract Pricing Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sign Into View Organizational & Contract Pricing Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur™, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sign Into View Organizational & Contract Pricing Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Sign Into View Organizational & Contract Pricing Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Sign Into View Organizational & Contract Pricing Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Sign Into View Organizational & Contract Pricing