MilliporeSigma
  • Chemical profile and biological activities of Veronica thymoides subsp. pseudocinerea.
Chemical profile and biological activities of Veronica thymoides subsp. pseudocinerea.
Pharmaceutical biology (2014-10-22)
Abdulselam Ertas, Mehmet Boga, Murat Kizil, Bircan Ceken, Ahmet Ceyhan Goren, Nesrin Hasimi, Serpil Demirci, Gulacti Topcu, Ufuk Kolak
ABSTRACT
In Turkey, Veronica species (Plantaginaceae) have been used as a diuretic and for wound healing in traditional medicine. To examine the fatty acid and essential oil profiles, the antioxidant, anticholinesterase, antimicrobial, and DNA damage effects of Veronica thymoides P.H. Davis subsp. pseudocinerea M.A. Fischer as a potential source of natural active compounds. GC/MS was used to analyze essential oil and fatty acid obtained from whole plant. The antioxidant activity was evaluated by the β-carotene-linoleic acid test system, DPPH-free and ABTS cation radicals scavenging, and cupric reducing antioxidant capacity assays. The anticholinesterase and antimicrobial activities were determined by Ellman and broth macrodillution methods, respectively. The effect of the methanol extract on DNA cleavage was investigated. Hexatriacontene (21.0%) was found to be the main constituent in essential oil, and linoleic acid (25.2%) and palmitic acid (20.6%) in fatty acid. Methanol extract demonstrated the best IC50 values in lipid peroxidation (49.81 ± 0.31 µg/ml) and DPPH-free radical scavenging activity (15.32 ± 0.17 µg/ml). Methanol and water extracts possessed strong ABTS cation radical scavenging activity with IC50 values 9.15 ± 0.28 and 8.90 ± 0.1  µg/ml, respectively. The acetone extract exhibited moderate butyrylcholinesterase inhibitory activity. The highest antimicrobial activity was determined in methanol extract against Escherichia coli with 31.25 µg/ml MIC value. Inhibition of methanol extract on plasmid DNA cleavage by OH radicals was found to be 93.32% at 500 µg/ml. The methanol extract having strong antioxidant and DNA damage effects could be investigated phytochemically to find natural active compounds.
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Diethyl ether, for residue analysis, JIS 5000
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%