MilliporeSigma
  • HJP272, a novel endothelin receptor antagonist, attenuates lipopolysaccharide-induced acute lung injury in hamsters.
HJP272, a novel endothelin receptor antagonist, attenuates lipopolysaccharide-induced acute lung injury in hamsters.
Lung (2014-08-05)
Shikha Patel, Xingjian Liu, Ming Liu, Ralph Stephani, Hardik Patel, Jerome Cantor
ABSTRACT
Previous studies from this laboratory indicate that endothelin-1 (ET-1), a potent vasoconstrictor, may play an important role in lipopolysaccharide (LPS)-induced release of neutrophils from the pulmonary microvasculature. To further test this concept, Syrian hamsters were treated with a novel endothelin receptor A (ETA) antagonist (HJP272) prior to intratracheal instillation of LPS. The effect of HJP272 on the LPS-induced inflammatory reaction was determined by measuring: (1) lung histopathological changes, (2) total neutrophils in bronchoalveolar lavage fluid (BALF), (3) expression of tumor necrosis factor receptor 1 (TNFR1) by BALF macrophages, and (4) alveolar septal cell apoptosis. Treatment with HJP272 significantly reduced each of these parameters during a 24-hr period following LPS instillation, supporting the concept that limiting the activity of ET-1 may reduce the extent of lung injury. This hypothesis was further tested by giving ET-1 prior to LPS instillation, which resulted in a marked enhancement of LPS-induced lung inflammation, as measured by BALF neutrophils and TNFR1-positive macrophages. Furthermore, the increase in neutrophils resulting from treatment with ET-1 was significantly reduced by HJP272, again demonstrating the ability of ETA receptor antagonists to limit the influx of these cells into the lung. These findings suggest a potential therapeutic role for these agents in diseases where neutrophils are a significant cause of lung injury, such as bronchopneumonia, respiratory distress syndrome, and chronic obstructive pulmonary disease.
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7