MilliporeSigma
  • Quantitative profiling of perfluoroalkyl substances by ultrahigh-performance liquid chromatography and hybrid quadrupole time-of-flight mass spectrometry.
Quantitative profiling of perfluoroalkyl substances by ultrahigh-performance liquid chromatography and hybrid quadrupole time-of-flight mass spectrometry.
Analytical and bioanalytical chemistry (2015-01-31)
Yolanda Picó, Marinella Farré, Damià Barceló
ABSTRACT
The accurate determination of perfluoroalkyl substances (PFSAs) in water, sediment, fish, meat, and human milk was achieved by ultrahigh-performance liquid chromatography-quadrupole time-of-flight mass spectrometry (UHPLC-QqTOF-MS) with an ABSciex Triple TOF®. A group of 21 PFSAs was selected as target to evaluate the quantitative possibilities. Full scan MS acquisition data allows quantification at relevant low levels (0.1-50 ng L(-1) in water, 0.05-2 ng g(-1) in sediments, 0.01-5 ng g(-1) in fish and meat, and 0.005-2 ng g(-1) in human milk depending on the compound). Automatic information dependent acquisition product ion mass spectrometry (IDA-MS/MS) confirms the identity even for those compounds that presented only one product ion. The preparation of a homemade database using the extracted ion chromatogram (XIC) Manager of the software based upon retention time, accurate mass, isotopic pattern, and MS/MS library searching achieves not only the successful identification of PFSAs but also of some pharmaceuticals, such as acetaminophen, ibuprofen, salicylic acid, and gemfibrozid. Mean recoveries and relative standard deviation (RSD) were 67-99% (9-16% RSD) for water, 62-103% (8-18% RSD) for sediment, 60-95% (8-17% RSD) for fish, 64-95% (8-15% RSD) for meat, and 63-95% (8-16%) for human milk. The quantitative data obtained for 60 samples by UHPLC-QqTOF-MS agree with those obtained by LC-MS/MS with a triple quadrupole (QqQ).
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Perfluorodecanoic acid, 98%
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Perfluoropentanoic acid, 97%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%