MilliporeSigma
Light-induced crawling of crystals on a glass surface.
Nature communications (2015-06-19)
Emi Uchida, Reiko Azumi, Yasuo Norikane
ABSTRACT
Motion is an essential process for many living organisms and for artificial robots and machines. To date, creating self-propelled motion in nano-to-macroscopic-sized objects has been a challenging issue for scientists. Herein, we report the directional and continuous motion of crystals on a glass surface when irradiated simultaneously with two different wavelengths, using simple azobenzenes as a photoresponsive organic compound. The direction of the motion can be controlled by the position of the light sources, and the crystals can even climb vertical surfaces. The motion is driven by crystallization and melting at the front and rear edges of the crystal, respectively, via photochemical conversion between the crystal and liquid phases induced by the trans-cis isomerization of azobenzenes. This finding could lead to remote-controlled micrometre-sized vehicles and valves on solid substrates.
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, ACS reagent, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
Pricing and availability is not currently available.
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, puriss. p.a., reag. Ph. Eur., ≥99% (perchloric acid titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Pricing and availability is not currently available.