MilliporeSigma
Transient and quasi-permanent networks in xyloglucan solutions.
Carbohydrate polymers (2015-06-09)
Rilton Alves de Freitas, Vivian C Spier, Maria Rita Sierakowski, Taco Nicolai, Lazhar Benyahia, Christophe Chassenieux
ABSTRACT
Viscoelastic properties of aqueous solutions of xyloglucan extracted from Hymenaea courbaril seeds (Jatobá gum) were investigated by rheology over a wide range of concentrations and temperatures. The polymer was characterized in dilute solutions by light scattering measurements and size exclusion chromatography. Xyloglucan formed, in semi-dilute solutions (C 0.3 wt%), a transient network with cross-links characterized by a broad distribution of lifetimes, independent of the temperature and concentration. Progressively, at higher temperatures (>60°C), a second much weaker quasi-permanent network was formed and attributed to the exchange of intra- to inter-chain bonds. The stiffness of the second network increased with decreasing temperature, but it could be easily broken by applying a relatively weak shear stress and is readily reversible on re-heating, and partially reversible on resting at 20°C.
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O