MilliporeSigma
  • Experimental and theoretical studies of spectroscopic properties of simple symmetrically substituted diphenylbuta-1,3-diyne derivatives.
Experimental and theoretical studies of spectroscopic properties of simple symmetrically substituted diphenylbuta-1,3-diyne derivatives.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2015-11-05)
Małgorzata Wierzbicka, Irena Bylińska, Artur Sikorski, Cezary Czaplewski, Wiesław Wiczk
ABSTRACT
A series of symmetrically substituted diphenylbuta-1,3-diyne (DPB) derivatives possessing electron-donating (N,N-dimethylamino or methoxy) or electron-accepting (nitrile, ester or aldehyde) groups have been prepared and studied with emphasis on their spectral and photophysical properties. The photophysical characteristics of these compounds have been studied in relation to their structures and influence of solvents or temperature. The observed spectral and photophysical properties are explained with the help of potential energy maps in the ground and excited states obtained from density functional theory (DFT, B3LYP, def2TZVP basis set) calculations. The structure-property relationship of all of the compounds is discussed and compared with appropriate diphenylacetylene derivatives.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
2-Methyl-3-butyn-2-ol, 98%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS