MilliporeSigma
  • Dehydrogenative TEMPO-Mediated Formation of Unstable Nitrones: Easy Access to N-Carbamoyl Isoxazolines.
Dehydrogenative TEMPO-Mediated Formation of Unstable Nitrones: Easy Access to N-Carbamoyl Isoxazolines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-28)
Andrea Gini, Marwin Segler, Dominik Kellner, Olga García Mancheño
ABSTRACT
N-carbamoyl nitrones represent an important class of reagents for the synthesis of a variety of natural and biologically active compounds. These compounds are generally converted into valuable 4-isoxazolines upon cyclization reaction with dipolarophiles. However, these types of N-protected nitrones are highly unstable, which limits their synthesis, storage and practical use, enforcing alternative lengthy or elaborated synthetic routes. In this work, a 2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO)-mediated formal "dehydrogenation" of N-protected benzyl-, allyl- and alkyl-substituted hydroxylamines followed by in situ trapping of the generated unstable nitrones into N-carbamoyl 4-isoxazolines is presented. A plausible mechanism is also proposed, in which the dipolarophile shows an important assistant role in the generation of the active nitrone intermediate. This simple protocol avoids the problematic isolation of N-carbamoyl protected nitrones, providing new synthetic possibilities in isoxazoline chemistry.
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Dimethyl acetylenedicarboxylate, 95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles