MilliporeSigma
  • Remyelination After Cuprizone-Induced Demyelination Is Accelerated in Juvenile Mice.
Remyelination After Cuprizone-Induced Demyelination Is Accelerated in Juvenile Mice.
Journal of neuropathology and experimental neurology (2015-06-27)
Sabine Pfeifenbring, Stefan Nessler, Christiane Wegner, Christine Stadelmann, Wolfgang Brück
ABSTRACT
Remyelination capacity decreases with age in adult mice, but data comparing remyelination capacity after toxic demyelination in developing mice versus adult mice are not available. We treated 3-week-old and adult C57BL/6 mice with cuprizone for 1 to 5 weeks and studied demyelination/remyelination and cellular reactions in the corpus callosum and motor cortex by histology, immunohistochemistry, and electron microscopy. We compared results between the 2 treated groups and age-matched controls. In juvenile mice, significant demyelination was detectable in the corpus callosum on Week 2 and in the motor cortex on Week 5. Oligodendrocyte loss, microglial activation, and acute axonal damage peaked on Week 2. Increased numbers of oligodendrocyte precursor cells were evident on Week 1, and remyelination was detectable on Week 3. Juvenile mice showed more rapid demyelination than adult mice, which may be related to greater vulnerability of oligodendrocytes, lower myelin content, or dose-dependent cuprizone effects. Earlier activation of microglia and proliferation of oligodendrocyte precursor cells probably contributed to accelerated remyelination and less pronounced axonal damage. Our data indicate that oligodendroglial regeneration and remyelination are enhanced in the maturing rodent brain compared with the young-adult rodent brain.
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)