MilliporeSigma
  • A plasticizer index to universally correlate the normalized work of fracture and elastic modulus of plasticized cellulose triacetates.
A plasticizer index to universally correlate the normalized work of fracture and elastic modulus of plasticized cellulose triacetates.
Carbohydrate polymers (2015-06-17)
Farzaneh Azadimanesh, Naser Mohammadi
ABSTRACT
Cellulose triacetate (CTA) plasticization with 25 wt% glycerin (GLY), triethyl citrate (TEC) or dioctyl phthalate (DOP) enhanced its crystalline content as much as 55%, while only 10% crystalline content increase noticed using poly (ethylene glycol) (PEG). In addition, GLY and PEG did not cause any sensible Tg depression regarding CTA, whereas DOP and TEC decreased it as much as 46°C and 16°C, respectively. Furthermore, the work of fracture (WoF) of CTA was deteriorated by PEG, did not change using GLY and magnified as much as 6.5 and 15 times by TEC and DOP, respectively. The elastic modulus (EM) of the plasticized CTAs, however, were 1.07, 1.06, 0.42 and 0.3 GPa using GLY, PEG, TEC and DOP, respectively. Finally, the normalized WoF and EM of the plasticized CTAs by their Tg depression (ΔTg) and crystalline content (Xc) showed a universal correlation with a plasticizer index containing its molar volume, concentration and polymer/plasticizer Painter-Coleman interaction parameter.
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Bis(2-ethylhexyl) phthalate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)
Sigma-Aldrich
Ethylene glycol 5 M solution