MilliporeSigma
  • Polymer-immobilized ready-to-use recombinant yeast assays for the detection of endocrine disruptive compounds.
Polymer-immobilized ready-to-use recombinant yeast assays for the detection of endocrine disruptive compounds.
Chemosphere (2015-03-24)
Michal Bittner, Sergio Jarque, Klára Hilscherová
ABSTRACT
Recombinant yeast assays (RYAs) constitute a suitable tool for the environmental monitoring of compounds with endocrine disrupting activities, notably estrogenicity and androgenicity. Conventional procedures require yeast reconstitution from frozen stock, which usually takes several days and demands additional equipment. With the aim of applying such assays to field studies and making them more accessible to less well-equipped laboratories, we have optimized RYA by the immobilization of Saccharomyces cerevisiae cells in three different polymer matrices - gelatin, Bacto agar, and Yeast Extract Peptone Dextrose agar - to obtain a ready-to-use version for the fast assessment of estrogenic and androgenic potencies of compounds and environmental samples. Among the three matrices, gelatin showed the best results for both testosterone (androgen receptor yeast strain; AR-RYA) and 17β-estradiol (estrogen receptor yeast strain; ER-RYA). AR-RYA was characterized by a lowest observed effect concentration (LOEC), EC50 and induction factor (IF) of 1nM, 2.2nM and 51, respectively. The values characterizing ER-RYA were 0.4nM, 1.8nM, and 63, respectively. Gelatin immobilization retained yeast viability and sensitivity for more than 90d of storage at 4°C. The use of the immobilized yeast reduced the assay duration to only 3h without necessity of sterile conditions. Because immobilized RYA can be performed either in multiwell microplates or glass tubes, it allows multiple samples to be tested at once, and easy adaptation to existing portable devices for direct in-field applications.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
YPD Agar
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Citric acid monohydrate, JIS special grade, ≥99.5%
Sigma-Aldrich
Citric acid monohydrate, ≥99.5%, suitable for amino acid analysis
Sigma-Aldrich
Sodium citrate tribasic dihydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Citric acid monohydrate, SAJ first grade, ≥99.5%
Sigma-Aldrich
Citric acid monohydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
Sigma-Aldrich
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ≥98%
Sigma-Aldrich
Citric acid monohydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Citric acid monohydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology