MilliporeSigma
  • Surfactant toxicity in a case of (4-chloro-2-methylphenoxy) acetic acid herbicide intoxication.
Surfactant toxicity in a case of (4-chloro-2-methylphenoxy) acetic acid herbicide intoxication.
Human & experimental toxicology (2014-11-16)
I Hwang, J W Lee, J S Kim, H W Gil, H Y Song, S Y Hong
ABSTRACT
Self-poisoning with (4-chloro-2-methylphenoxy) acetic acid (MCPA) is a common reason for presentation to hospitals, especially in some Asian countries. We encountered a case of a 76-year-old woman who experienced unconsciousness, shock and respiratory failure after ingesting 100 mL MCPA herbicide. We determined whether the surfactant in the formulation was the chemical responsible for the toxic symptom in this patient. 3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) cell viability and lactate dehydrogenase (LDH) cytotoxicity assays were performed on human brain neuroblastoma SK-N-SH cells. The expressions of 84 genes in 9 categories that are implicated in cellular damage pathways were quantified using an RT(2) Profiler™ PCR array on a human neuronal cell line challenged with polyoxyethylene tridecyl ether (PTE). Pesticide intoxication institute in university hospital. Extracorporeal elimination with intravenous lipid emulsion. Cell viability and gene expression. In the MTT assay, MCPA only minimally decreased cell viability even at concentrations as high as 1 mM. Cells treated with 1-methoxy-2-propanol, dimethylamine and polypropylene glycol exhibited minimal decreases in viability, whilst the viability of cells challenged with PTE decreased dramatically; only 15.5% of cells survived after exposure to 1 µM PTE. Similarly, the results of the LDH cytotoxicity assay showed that MCPA had very low cytotoxicity, whilst cells treated with PTE showed incomparably higher LDH levels (p < 0.0001). PTE up-regulated the expressions of genes implicated in various cell damage pathways, particularly genes involved in the inflammatory pathway. The surfactant PTE was likely the chemical responsible for the toxic symptom in our patient.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
4-Chloro-2-methylphenoxyacetic acid, technical, ≥95.0% (T)
Sigma-Aldrich
Polypropylene glycol, P 2,000
Sigma-Aldrich
Polypropylene glycol, P 400
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Poly(propylene glycol), average Mn ~725
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,700
Sigma-Aldrich
Poly(propylene glycol), average Mn ~1,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~4,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~425
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethylamine solution, CP, 50% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis