MilliporeSigma
  • Copper, selenium and zinc levels after bariatric surgery in patients recommended to take multivitamin-mineral supplementation.
Copper, selenium and zinc levels after bariatric surgery in patients recommended to take multivitamin-mineral supplementation.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS) (2014-10-02)
Dimitris Papamargaritis, Erlend T Aasheim, Barry Sampson, Carel W le Roux
ABSTRACT
Bariatric surgery is widely performed to improve obesity-related disorders, but can lead to nutrient deficiencies. In this study we examined serum trace element concentrations before and after bariatric surgery. We obtained serum trace element concentrations by inductively coupled plasma-mass spectrometry (ICP-MS) method in 437 patients (82% women, median preoperative body-mass index 46.7 kg/m(2) [interquartile range 42-51]) undergoing either gastric banding (22.7%), sleeve gastrectomy (20.1%), or gastric bypass (57.3%) procedures. Trace element data were available for patients preoperatively (n = 44); and 3 (n = 208), 6 (n = 174), 12 (n = 122), 18 (n = 39), 24 (n = 44) and 36 months (n = 14) post-operatively. All patients were recommended to take a multivitamin-mineral supplement after surgery. Copper deficiency was found in 2% of patients before surgery; and after surgery deficiency rates ranged from 0 to 5% with no significant change in median concentrations during follow-up (p = 0.68). Selenium deficiency was reported in 2% of patients before surgery; and after surgery deficiency rates ranged from 11 to 15% with a near-significant change in median concentrations (p = 0.056). Zinc deficiency was reported in 7% before surgery; and after surgery deficiency rates ranged from 7 to 15% with no significant change in median concentrations (p = 0.39). In bariatric surgery patients recommended to take multivitamin-mineral supplements, serum copper, zinc and selenium concentrations were mostly stable during the first years after bariatric surgery. There was a possible tendency for selenium concentrations to decline during the early postoperative period.
Product Description
Supelco
1-Butanol, suitable for HPLC, 99.8%
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Butanol, suitable for HPLC
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG