MilliporeSigma
  • The in situ gas-phase formation of a C-glycoside ion obtained during electrospray ionization tandem mass spectrometry. A unique intramolecular mechanism involving an ion-molecule reaction.
The in situ gas-phase formation of a C-glycoside ion obtained during electrospray ionization tandem mass spectrometry. A unique intramolecular mechanism involving an ion-molecule reaction.
Rapid communications in mass spectrometry : RCM (2015-09-04)
Joseph H Banoub, Wael L L Demian, Paolo Piazzetta, George Sarkis, Basem Kanawati, Dominique Lafont, Nicolas Laurent, Celine Vaillant, Edward Randell, Gianluca Giorgi, Travis D Fridgen
ABSTRACT
This study examines the electrospray ionization mass spectrometry (ESI-MS), in-source collision-induced dissociation (CID) fragmentation and low-energy collision-induced dissociation tandem mass spectrometry (CID-MS/MS) of a synthetic pair of β- and α-anomers of the amphiphilic cholesteryl polyethoxy neoglycolipids containing the 2-azido-2-deoxy-D-galactosyl-D-GalN3 moiety. We describe the novel and unique in situ gas-phase formation of a C-glycoside ion formed during all these gas-phase processes and propose a reasonable mechanism for its formation. The synthetic amphiphilic glycolipids were composed of the 2-deoxy-2-azido-D-galactosyl moiety (GalN3, the hydrophilic part) covalently attached to a polyethoxy spacer which is covalently linked to the cholesteryl moiety (hydrophobic part). The 2-azido-2-deoxy-α- and β-D-galactosyl-containing glycolipids were studied by in-time and in-space ESI-MS and CID-MS/MS in positive ion mode, with quadrupole ion trap (QIT), quadrupole-quadrupole-time-of-flight (QqTOF), and Fourier transform ion cyclotron resonance (FTICR) instruments. Conventional single-stage ESI-MS analysis showed the formation of the protonated molecule. During the single-stage ESI-MS analysis and the CID-MS/MS of the [M+H](+) and [M+NH4](+) adducts obtained from both glycolipid anomers, the presence of a series of specific product ions with different intensities was observed, consistent with the [C-glycoside+H-N2](+), [cholestadiene+H](+), 2-deoxy-2-D-azido-galactosyl [GalN3](+), [GalNH](+) and [sugar-Spacer+H](+) ions. The gas-phase formation of the [C-glycoside+H-N2](+) ion isolated from the glycolipid anomers was observed during both the ESI-MS of the glycolipids and the CID-MS/MS analyses of the [M+H](+) ions and it was found to occur by an intramolecular rearrangement involving an ion-molecule complex. CID-QqTOF-MS/MS and CID-FTICR-MS(2) analysis allowed the differentiation of the two glycolipid anomers and showed noticeable variation in the intensities of the product ions.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 23, 2025
$27.00
500 mL
Estimated to ship on March 23, 2025
$73.50
1 L
In Stock
Details...
$137.00
Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 vial
Estimated to ship on March 23, 2025
$37.80
5 vials
Estimated to ship on March 23, 2025
$131.00
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
$27.00
$22.95
500 mL
In Stock
Details...
$61.60
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 L
In Stock
Details...
$76.60
10 x 50 mL
In Stock
Details...
$129.00
6 x 1 L
Estimated to ship on March 23, 2025
$377.00
10 L
In Stock
Details...
$392.00
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
$16.20
500 mL
In Stock
Details...
$22.20
1 L
In Stock
Details...
$44.80
6 x 500 mL
Estimated to ship on March 23, 2025
$117.00
6 x 1 L
Estimated to ship on March 23, 2025
$204.00
24 x 500 mL
Estimated to ship on March 23, 2025
$409.00
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
$65.40
500 mL
In Stock
Details...
$112.00
1 L
In Stock
Details...
$242.00
6 x 100 mL
In Stock
Details...
$304.00
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mL
In Stock
Details...
$53.50
$48.15
30 mL
Estimated to ship on March 23, 2025
$75.60
Sigma-Aldrich
Water, BioPerformance Certified
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Estimated to ship on March 23, 2025
$23.30
1 L
In Stock
Details...
$88.90
Sigma-Aldrich
Methanol, NMR reference standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 pkg
Estimated to ship on March 23, 2025
$188.00
1 pkg
Estimated to ship on March 23, 2025
$193.00
$38.60
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
In Stock
Details...
$224.00
$156.80
4 x 4 L
In Stock
Details...
$752.00
$639.20
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ special grade
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 pkg
Estimated to ship on March 23, 2025
$281.00
$56.20
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
In Stock
Details...
$54.60
1 kg
In Stock
Details...
$91.80
5 kg
In Stock
Details...
$357.00
$317.73
10 kg
In Stock
Details...
$510.00
25 kg
In Stock
Details...
$1,030.00
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 x 4 L
In Stock
Details...
$1,230.00