MilliporeSigma
  • Immobilisation of a molecular epoxidation catalyst on UiO-66 and -67: the effect of pore size on catalyst activity and recycling.
Immobilisation of a molecular epoxidation catalyst on UiO-66 and -67: the effect of pore size on catalyst activity and recycling.
Dalton transactions (Cambridge, England : 2003) (2015-08-19)
Marlene Kaposi, Mirza Cokoja, Christine H Hutterer, Simone A Hauser, Tobias Kaposi, Florian Klappenberger, Alexander Pöthig, Johannes V Barth, Wolfgang A Herrmann, Fritz E Kühn
ABSTRACT
Amino-functionalised metal-organic frameworks UiO-66 and -67 were post-synthetically modified with salicylaldehyde. A molybdenum complex was immobilised on the resulting materials. They were characterised by (13)C-MAS-NMR, XPS and PXRD to confirm immobilisation and stability. The immobilised complex is an active and reusable catalyst for olefin epoxidation with tert-butyl hydroperoxide (TBHP) as an oxidant. It is shown that the effective pore size, probed with Brunauer-Emmett-Teller (BET) surface area analysis and the number of amino groups affect the diffusion of reactants and products, as well as catalyst recycling.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, for molecular biology, sterile filtered
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
E-Toxate Water, endotoxin, free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Water, BioPerformance Certified