MilliporeSigma
  • Lycium intricatum Boiss.: An unexploited and rich source of unsaturated fatty acids, 4-desmethylsterols and other valuable phytochemicals.
Lycium intricatum Boiss.: An unexploited and rich source of unsaturated fatty acids, 4-desmethylsterols and other valuable phytochemicals.
Lipids in health and disease (2015-06-25)
Abdennacer Boulila, Afef Bejaoui
ABSTRACT
Lycium intricatum Boiss., a Solanaceous shrubbery is used in Tunisia as a windbreak and medicinal plant. However, it is considered as underexploited specie despite its high potential to serve as source with economic and nutritional value. To date only limited information about its phytochemistry, especially of its oil has been published. This work provides data on fatty acids, phytosterols and vitamin D composition of L. intricatum seed oil. It opens up new possibilities of developing L. intricatum as a new crop that contains phytochemicals with high added value little influenced by selection or commercial breeding. The composition of fatty acids, phytosterols and vitamin D in L. intricatum seed oil was assessed by GC-FID. The main fatty acids of L. intricatum seed oil were linoleic acid (49.47%), palmitoleic acid (27.96%) and erucic acid (13.62%). Palimtic acid was present at low percentage (0.63%). The content of unsaturated fatty acids was high as 94.04%. The sterolic fraction was composed of stigmasterol (18.56 mg/100 g), β-sitosterol (13.04 mg/100 g). L. intricatum oil is an oily matrix that contains hydrocarbons, mainly squalene (63.36 mg/100 g), and two triterpenic alcohol erythrodiol (80.36 mg/100 g) and uvaol (24.06 mg/100 g). provitamin D was present in high quantity (8.12 mg/100 g). From these results it has been shown that L. intricatum seeds have great potential as a source of fatty acids and phytosterols for natural health products.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Sodium sulfate, JIS special grade, ≥99.0%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)