MilliporeSigma
  • Fibroblast growth factor 21 prevents atherosclerosis by suppression of hepatic sterol regulatory element-binding protein-2 and induction of adiponectin in mice.
Fibroblast growth factor 21 prevents atherosclerosis by suppression of hepatic sterol regulatory element-binding protein-2 and induction of adiponectin in mice.
Circulation (2015-03-22)
Zhuofeng Lin, Xuebo Pan, Fan Wu, Dewei Ye, Yi Zhang, Yu Wang, Leigang Jin, Qizhou Lian, Yu Huang, Hong Ding, Chris Triggle, Kai Wang, Xiaokun Li, Aimin Xu
ABSTRACT
Fibroblast growth factor 21 (FGF21) is a metabolic hormone with pleiotropic effects on glucose and lipid metabolism and insulin sensitivity. It acts as a key downstream target of both peroxisome proliferator-activated receptor α and γ, the agonists of which have been used for lipid lowering and insulin sensitization, respectively. However, the role of FGF21 in the cardiovascular system remains elusive. The roles of FGF21 in atherosclerosis were investigated by evaluating the impact of FGF21 deficiency and replenishment with recombinant FGF21 in apolipoprotein E(-/-) mice. FGF21 deficiency causes a marked exacerbation of atherosclerotic plaque formation and premature death in apolipoprotein E(-/-) mice, which is accompanied by hypoadiponectinemia and severe hypercholesterolemia. Replenishment of FGF21 protects against atherosclerosis in apolipoprotein E(-/-)mice via 2 independent mechanisms, inducing the adipocyte production of adiponectin, which in turn acts on the blood vessels to inhibit neointima formation and macrophage inflammation, and suppressing the hepatic expression of the transcription factor sterol regulatory element-binding protein-2, thereby leading to reduced cholesterol synthesis and attenuation of hypercholesterolemia. Chronic treatment with adiponectin partially reverses atherosclerosis without obvious effects on hypercholesterolemia in FGF21-deficient apolipoprotein E(-/-) mice. By contrast, the cholesterol-lowering effects of FGF21 are abrogated by hepatic expression of sterol regulatory element-binding protein-2. FGF21 protects against atherosclerosis via fine tuning the multiorgan crosstalk among liver, adipose tissue, and blood vessels.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
SKU
Pack Size
Availability
Price
Quantity
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
SAFC
Cholesterol, Plant-Derived, SyntheChol®