MilliporeSigma
  • Quantification of arginine and its methylated derivatives in healthy children by liquid chromatography-tandem mass spectrometry.
Quantification of arginine and its methylated derivatives in healthy children by liquid chromatography-tandem mass spectrometry.
Journal of chromatographic science (2014-09-28)
Fernando Andrade, Marta Llarena, Sergio Lage, Luis Aldámiz-Echevarría
ABSTRACT
Asymmetric dimethylarginine (ADMA) is a competitive inhibitor of nitric oxide synthase, which is responsible for most of the vascular nitric oxide (NO) produced. NO is an important physiological mediator of vascular tone and structure in normally functioning endothelial cells. We report the optimization of a liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for simultaneous determination of arginine (Arg) and its derivatives in biological samples. Chromatographic separation and mass detection were performed by reverse phase chromatography coupled with tandem mass spectrometry. For sample preparation, plasma proteins were removed by centrifugal filters. Positive electrospray ionization was performed and analytes were detected by multiple reaction monitoring. Inter- and intra-day repeatability, accuracy, recovery, and limits of detection and quantification were evaluated to validate the method. Plasma and urine levels were measured in healthy children to establish control values: 52.2-124.7 µM for Arg, 0.06-0.16 µM for MMA, 0.42-1.10 µM for ADMA and 0.41-0.96 µM for SDMA in plasma. Quantification of Arg and its methylated derivatives by LC-MS/MS can be carried out without the need of organic solvents for sample preparation, and be used as a valuable tool in research on endothelial dysfunction.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride