MilliporeSigma
  • Upregulation of circulating components of the alternative renin-angiotensin system in inflammatory bowel disease: A pilot study.
Upregulation of circulating components of the alternative renin-angiotensin system in inflammatory bowel disease: A pilot study.
Journal of the renin-angiotensin-aldosterone system : JRAAS (2014-02-08)
Mayur Garg, Louise M Burrell, Elena Velkoska, Karen Griggs, Peter W Angus, Peter R Gibson, John S Lubel
ABSTRACT
The relationship between intestinal inflammation and circulating components of the renin-angiotensin system (RAS) is poorly understood. Demographic and clinical data were obtained from healthy controls and patients with inflammatory bowel disease (IBD). Plasma concentrations of the classical RAS components (angiotensin-converting enzyme (ACE) and angiotensin II (Ang II)) and alternative RAS components (ACE2 and angiotensin (1-7) (Ang (1-7))) were analysed by radioimmuno- and enzymatic assays. Systemic inflammation was assessed using serum C-reactive protein (CRP), white cell count, platelet count and albumin, and intestinal inflammation by faecal calprotectin. Nineteen healthy controls (11 female; mean age 38 years, range 23-68), 19 patients with Crohn's disease (11 female; aged 45 years, range 23-76) and 15 patients with ulcerative colitis (6 female; aged 42 years, 26-64) were studied. Circulating classical RAS component levels were similar across the three groups, whereas ACE2 activity and Ang (1-7) concentrations were higher in patients with IBD compared to controls (ACE2: 21.5 vs 13.3 pmol/ml/min, p<0.05; Ang (1-7): 22.8 vs 14.1 pg/ml, p<0.001). Ang (1-7) correlated weakly with platelet and white cell counts, but not calprotectin or CRP, in patients with IBD. Circulating components of the alternative RAS are increased in patients with IBD.
Sigma-Aldrich
6-Maleimidohexanoic acid, 90% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$36.30
500 g
In Stock
Details...
$55.30
3 kg
In Stock
Details...
$196.00
12 kg
In Stock
Details...
$541.00
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
In Stock
Details...
$36.30
500 g
In Stock
Details...
$55.30
3 kg
In Stock
Details...
$196.00
12 kg
In Stock
Details...
$541.00