MilliporeSigma
  • Structural and electronic characterization of multi-electron reduced naphthalene (BIAN) cobaloximes.
Structural and electronic characterization of multi-electron reduced naphthalene (BIAN) cobaloximes.
Dalton transactions (Cambridge, England : 2003) (2015-04-14)
Owen M Williams, Alan H Cowley, Michael J Rose
ABSTRACT
Reported here are the syntheses and characterization of cobaloximes that feature a bis(imino)acenaphthene (BIAN) appended ligand. The X-ray crystal structures and spectroscopy ((1)H NMR or EPR) of the complexes within the series [Co(aqdBF2)2(MeCN)2] (), [Co(aqdBF2)(MeCN)2](-) () and [Co(aqdBF2)2(MeCN)2](2-) (, ) are reported and the 3-electron reduced complex [Co(aqdBF2)2(MeCN)2](3-) () has been prepared in situ and characterized by (1)H NMR spectroscopy. The X-ray crystal structures revealed the presence of a 6-coordinate Co(II) species (), a 5-coordinate Co(I) species (), and a 4-coordinate complex (, ). In the case of complex , evidence from single crystal EPR spectroscopy (g‖ = 2.017, g⊥ = 1.987; <10 G linewidths) in conjunction with DFT calculations indicate that the EPR signal originates from a delocalized ligand-based unpaired spin. The frontier orbitals obtained from DFT calculations on , , , & support the electronic assignments that were observed spectroscopically. The cathodic cyclic voltammogram (CV) of the solvato congener in DMF solution, namely [Co(aqdBF2)2(DMF)2], exhibits three reversible redox events near -1.0, -1.5 and -2.0 V vs. Fc/Fc(+). Catalytic proton reduction was observed by CV near the third redox peak. Compared with other cobaloximes (Ecat = -1.0 V), the delay of catalytic onset arises from the existence of a series of resonance-stabilized intermediates.
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
12-Crown-4, 98%
Sigma-Aldrich
Boron trifluoride, electronic grade, ≥99.99%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
Mercury, SAJ first grade, ≥99.5%
Sigma-Aldrich
Mercury, JIS special grade, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Dibenzo-18-crown-6, 98%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Cobalt(II) acetate, 99.99% trace metals basis