MilliporeSigma
  • Molecular species identification, host preference and detection of myxoma virus in the Anopheles maculipennis complex (Diptera: Culicidae) in southern England, UK.
Molecular species identification, host preference and detection of myxoma virus in the Anopheles maculipennis complex (Diptera: Culicidae) in southern England, UK.
Parasites & vectors (2015-08-15)
Victor A Brugman, Luis M Hernández-Triana, Sean W J Prosser, Chris Weland, David G Westcott, Anthony R Fooks, Nicholas Johnson
ABSTRACT
Determining the host feeding patterns of mosquitoes by identifying the origin of their blood-meals is an important part of understanding the role of vector species in current and future disease transmission cycles. Collecting large numbers of blood-fed mosquitoes from the field is difficult, therefore it is important to maximise the information obtained from each specimen. This study aimed to use mosquito genome sequence to identify the species within Anopheles maculipennis sensu lato (An. maculipennis s.l.), identify the vertebrate hosts of field-caught blood-fed An. maculipennis s.l. , and to test for the presence of myxoma virus (Poxviridae, genus Leporipoxvirus) in specimens found to have fed on the European rabbit (Oryctolagus cuniculus). Blood-fed An. maculipennis s.l. were collected from resting sites at Elmley Nature Reserve, Kent, between June and September 2013. Hosts that An. maculipennis s.l. had fed on were determined by a PCR-sequencing approach based on the partial amplification of the mitochondrial cytochrome C oxidase subunit I gene. Mosquitoes were then identified to species by sequencing a region of the internal transcribed spacer-2. DNA extracts from all mosquitoes identified as having fed on rabbits were subsequently screened using PCR for the presence of myxoma virus. A total of 94 blood-fed Anopheles maculipennis s.l. were collected, of which 43 (46%) provided positive blood-meal identification results. Thirty-six of these specimens were identified as Anopheles atroparvus, which had fed on rabbit (n = 33, 92%) and cattle (n = 3, 8%). Seven mosquitoes were identified as Anopheles messeae, which had fed on cattle (n = 6, 86%) and dog (n = 1, 14%). Of the 33 An. atroparvus that contained rabbit blood, nine (27%) were positive for myxoma virus. Results demonstrate that a single DNA extract from a blood-fed mosquito can be successfully used for molecular identification of members of the An. maculipennis complex, blood-meal identification, and for the targeted detection of a myxoma virus. This study shows that An. atroparvus has a strong feeding preference for both healthy and myxoma-infected rabbits, providing evidence that this species may play a significant role in the transmission of myxomatosis among wild rabbit populations in the United Kingdom (UK).
Sigma-Aldrich
Water, BioPerformance Certified
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, PCR Reagent
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sign Into View Organizational & Contract Pricing
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sign Into View Organizational & Contract Pricing