MilliporeSigma
  • Rapid label-free determination of ketamine in whole blood using secondary ion mass spectrometry.
Rapid label-free determination of ketamine in whole blood using secondary ion mass spectrometry.
Talanta (2015-06-17)
Hua-Yang Liao, Jung-Hsuan Chen, Jing-Jong Shyue, Chia-Tung Shun, Huei-Wen Chen, Su-Wei Liao, Chih-Kang Hong, Pai-Shan Chen
ABSTRACT
A fast and accurate drug screening to identify the possible presence of a wide variety of pharmaceutical and illicit drugs is increasingly requested in forensic and clinical toxicology. The current first-line screening relies on immunoassays. They determine only certain common drugs of which antibodies are commercially available. To address the issue, a rapid screening using secondary ion mass spectrometry (SIMS) has been developed. In the study, SIMS directly analyzed ketamine in whole blood without any pretreatment. While the untreated blood has a complicated composition, principal-components analysis (PCA) is used to detect unknown specimens by building up an analytical model from blank samples which were spiked with ketamine at 100 ng mL(-1), to simulate the presence of ketamine. Each characteristics m/z is normalized and scaled by multiplying the root square of intensity and square of corresponding m/z, developed by National Institute of Standards and Technology (NIST). Using linear regression and the result of PCA, this study enables to correctly distinguish ketamine positive and negative groups in an unknown set of specimens. The quantity of ketamine in an unknown set was determined using gas chromatography-mass spectrometry (GC-MS) as the reference methodology. Instead limited by commercially available antibodies, SIMS detects target molecules straight despite the label-free detection capabilities of SIMS, additional data processing (here, PCA) can be used to fully analyse the produced data, which extends the range of analytes of interest on drug screening. Furthermore, extremely low sample volume, 5 µL, is required owing to the high spatial resolution of SIMS. In addition, while the whole blood is analyzed within 3 min, the whole analysis has been shortened significantly and high throughput can be achieved.
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexane, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexane, anhydrous, 95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.