MilliporeSigma
  • Hydrogen Sulfide Treatment Mitigates Renal Allograft Ischemia-Reperfusion Injury during Cold Storage and Improves Early Transplant Kidney Function and Survival Following Allogeneic Renal Transplantation.
Hydrogen Sulfide Treatment Mitigates Renal Allograft Ischemia-Reperfusion Injury during Cold Storage and Improves Early Transplant Kidney Function and Survival Following Allogeneic Renal Transplantation.
The Journal of urology (2015-08-05)
Ian Lobb, Michael Davison, David Carter, Weihua Liu, Aaron Haig, Lakshman Gunaratnam, Alp Sener
ABSTRACT
Ischemia-reperfusion injury is unavoidable during organ transplantation. Prolonged ischemia-reperfusion injury is detrimental to short-term and long-term graft function and survival. H2S is a recently characterized, endogenously produced gaseous molecule with important physiological roles that has been shown to be cytoprotective during tissue ischemia-reperfusion injury. The current study aimed to determine whether H2S could mitigate cold renal ischemia-reperfusion injury in the clinically relevant context of allogeneic renal transplantation. Following bilateral native nephrectomy Lewis rats underwent renal transplantation with kidneys from Brown Norway donor rats that were flushed with cold (4C) standard University of Wisconsin preservation solution (University of Wisconsin preservation solution group) or cold University of Wisconsin preservation solution plus 150 μM NaHS (H2S group) solution. Kidneys were stored for 6 hours at 4C in the same solution. Recipient animals were monitored for 14 days or until sacrifice using metabolic cages to assess various parameters of renal graft function. H2S treatment improved early allograft survival and function, and decreased early levels of necrosis, apoptosis and Kim-1 compared to University of Wisconsin preservation solution alone. H2S treatment did not affect allograft rejection. Rather, it modulated the early allograft transcriptome to decrease the expression of renal injury, coagulation and cellular stress response genes, and increase the expression of cellular proliferation and Ifn-γ induced genes compared to University of Wisconsin preservation solution alone. To our knowledge our findings are the first to show that H2S protects donor kidneys against cold ischemia-reperfusion injury in the context of allogeneic renal transplantation. This potentially represents a novel cost-effective therapeutic solution to mitigate ischemia-reperfusion injury and improve the clinical outcomes of renal transplantation.
Sigma-Aldrich
Hydrogen sulfide, ≥99.5%
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Sulfur-32S, 99.9 atom % 32S
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF