MilliporeSigma
  • MCPIP1 down-regulates IL-2 expression through an ARE-independent pathway.
MCPIP1 down-regulates IL-2 expression through an ARE-independent pathway.
PloS one (2012-11-28)
Min Li, Wenqiang Cao, Haifeng Liu, Wei Zhang, Xia Liu, Zhijian Cai, Jing Guo, Xuelian Wang, Zhaoyuan Hui, Hang Zhang, Jianli Wang, Lie Wang
ABSTRACT
IL-2 plays a key role in the survival and proliferation of immune cells, especially T lymphocytes. Its expression is precisely regulated at transcriptional and posttranscriptional level. IL-2 is known to be regulated by RNA binding proteins, such as tristetraprolin (TTP), via an AU-rich element (ARE) in the 3'-untranslated region (3'UTR) to influence the stability of mRNA. MCPIP1, identified as a novel RNase, can degrade IL-6, IL-12 and TNF-α mRNA by an ARE-independent pathway in the activation of macrophages. Here, we reported that MCPIP1 was induced in the activation of T lymphocytes and negatively regulated IL-2 gene expression in both mouse and human primary T lymphocytes through destabilizing its mRNA. A set of Luciferase reporter assay demonstrated that a non-ARE conserved element in IL-2 3'UTR, which formed a stem-loop structure, responded to MCPIP1 activity.RNA immunoprecipitation and Biotin pulldown experiments further suggested that MCPIP1 could modestly bind to IL-2 mRNA. Taken together, these data demonstrate that MCPIP1 down-regulates IL-2 via an ARE-independent pathway.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Roche
T7 RNA Polymerase, from Escherichia coli BL 21/pAR 1219