MilliporeSigma
The role of glucose 6-phosphate in the control of glycogen synthase.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (1997-06-01)
C Villar-Palasí, J J Guinovart
ABSTRACT
Elevated blood glucose concentrations result in increased intracellular levels of glucose 6-phosphate in liver, skeletal muscle, and adipose tissue. In liver, blood glucose concentrations are the main factor in control of the synthesis of glycogen; insulin has only a potentiating effect. In skeletal muscle and adipocytes, glucose alone has little effect on the activity of glycogen synthase, the limiting enzyme in glycogen synthesis. However, insulin released as a result of elevated blood glucose stimulates the translocation of specific glucose transporters to the cell membrane, increases the uptake of glucose, and causes the covalent, dephosphorylation-mediated activation of glycogen synthase. We present evidence that elevated intracellular contents of glucose 6-phosphate provoke the activation of glycogen synthase in liver, muscle, and adipose tissue. In addition, glucose 6-phosphate may inhibit the phosphorylation of glycogen synthase by cyclic AMP-stimulated protein kinase. We show that the stimulated glucose uptake and phosphorylation appear to play a major role in the control by insulin of the enzymes involved in glycogen synthesis.
Product Description
Roche
Glucose-6-phosphate, mol wt (glucose-6-P: Mr = 260.2, glucose-6-P-Na2: Mr = 304.2), pkg of 5 g