MilliporeSigma
  • Regulation of Ca2+ channels by SNAP-25 via recruitment of syntaxin-1 from plasma membrane clusters.
Regulation of Ca2+ channels by SNAP-25 via recruitment of syntaxin-1 from plasma membrane clusters.
Molecular biology of the cell (2016-11-02)
Trine Lisberg Toft-Bertelsen, Iwona Ziomkiewicz, Sébastien Houy, Paulo S Pinheiro, Jakob B Sørensen
ABSTRACT
SNAP-25 regulates Ca
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Sigma-Aldrich
Triton X-100, laboratory grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Sigma-Aldrich
Triton X-100, for molecular biology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00