MilliporeSigma
  • Phosphorylation of the deubiquitinase USP20 by protein kinase A regulates post-endocytic trafficking of β2 adrenergic receptors to autophagosomes during physiological stress.
Phosphorylation of the deubiquitinase USP20 by protein kinase A regulates post-endocytic trafficking of β2 adrenergic receptors to autophagosomes during physiological stress.
The Journal of biological chemistry (2015-02-11)
Reddy Peera Kommaddi, Pierre-Yves Jean-Charles, Sudha K Shenoy
ABSTRACT
Ubiquitination by the E3 ligase Nedd4 and deubiquitination by the deubiquitinases USP20 and USP33 have been shown to regulate the lysosomal trafficking and recycling of agonist-activated β2 adrenergic receptors (β2ARs). In this work, we demonstrate that, in cells subjected to physiological stress by nutrient starvation, agonist-activated ubiquitinated β2ARs traffic to autophagosomes to colocalize with the autophagy marker protein LC3-II. Furthermore, this trafficking is synchronized by dynamic posttranslational modifications of USP20 that, in turn, are induced in a β2AR-dependent manner. Upon β2AR activation, a specific isoform of the second messenger cAMP-dependent protein kinase A (PKAα) rapidly phosphorylates USP20 on serine 333 located in its unique insertion domain. This phosphorylation of USP20 correlates with a characteristic SDS-PAGE mobility shift of the protein, blocks its deubiquitinase activity, promotes its dissociation from the activated β2AR complex, and facilitates trafficking of the ubiquitinated β2AR to autophagosomes, which fuse with lysosomes to form autolysosomes where receptors are degraded. Dephosphorylation of USP20 has reciprocal effects and blocks trafficking of the β2AR to autophagosomes while promoting plasma membrane recycling of internalized β2ARs. Our findings reveal a dynamic regulation of USP20 by site-specific phosphorylation as well as the interdependence of signal transduction and trafficking pathways in balancing adrenergic stimulation and maintaining cellular homeostasis.
Sigma-Aldrich
(−)-Isoproterenol (+)-bitartrate salt, powder
Millipore
Protein G Plus/Protein A-Agarose, A mixture of Protein G PLUS and Protein A covalently conjugated to agarose. Useful for purification of IgG from biological fluids.
Sigma-Aldrich
Forskolin, Coleus forskohlii
Millipore
EZview Red Anti-HA Affinity Gel
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)