MilliporeSigma
  • Expression of the Na+/Ca2+ exchanger emerges in hepatic stellate cells after activation in association with liver fibrosis.
Expression of the Na+/Ca2+ exchanger emerges in hepatic stellate cells after activation in association with liver fibrosis.
Proceedings of the National Academy of Sciences of the United States of America (1998-06-06)
T Nakamura, S Arii, K Monden, M Furutani, Y Takeda, M Imamura, M Tominaga, Y Okada
ABSTRACT
Activation of hepatic stellate (Ito) cells is a final common pathway of liver fibrosis. The findings presented in this paper indicate that expression of Na+/Ca2+ exchanger (NCX) emerges in rat hepatic stellate cells after activation in vitro during primary culture or in vivo in response to intoxication with CCl4. NCX mRNA became detectable by Northern blot analysis in cultured stellate cells on day 3, as was alpha-smooth muscle actin, an indicator not only of smooth muscle differentiation but also of stellate cell activation. Western blot analysis showed expression of the exchanger protein in the activated stellate cells. Functional expression of the exchanger, monitored by Ni2+-sensitive, verapamil-insensitive intracellular free Ca2+ increases in response to reduction of extracellular Na+ concentration, became sizable by using Fura-2 in stellate cells by 7 days in culture. Furthermore, increased expression of the exchanger mRNA was found predominantly in stellate cells freshly isolated from the CCl4 model rat of hepatic fibrosis. Thus, it is concluded that NCX expression is closely associated with activation of hepatic stellate cells in vitro and in vivo. Because, even at the whole liver level, increased expression of NCX mRNA became observable after induction of liver fibrosis, it is suggested that NCX expression serves a useful diagnostic marker of liver fibrosis or cirrhosis.
Sigma-Aldrich
IgG2a−FITC Isotype Control from murine myeloma, clone UPC-10, purified immunoglobulin, buffered aqueous solution