Merck
  • Characterization of an immobilized enzyme reactor for on-line protein digestion.
Characterization of an immobilized enzyme reactor for on-line protein digestion.
Journal of chromatography. A (2016-11-24)
Stephanie Moore, Stephanie Hess, James Jorgenson
ABSTRACT
Despite the developments for faster liquid chromatographic and mass spectral detection techniques, the standard in-solution protein digestion for proteomic analyses has remained relatively unchanged. The typical in-solution trypsin protein digestion is usually the slowest part of the workflow, albeit one of the most important. The development of a highly efficient immobilized enzyme reactor (IMER) with rapid performance for on-line protein digestion would greatly decrease the analysis time involved in a proteomic workflow. Presented here is the development of a silica based IMER for on-line protein digestion, which produced rapid digestions in the presence of organic mobile phase for both model proteins and a complex sample consisting of the insoluble portion of a yeast cell lysate. Protein sequence coverage and identifications evaluated between the IMER and in-solution digestions were comparable. Overall, for a yeast cell lysate with only a 10s volumetric residence time on-column, the IMER identified 507 proteins while the in-solution digestion identified 490. There were no significant differences observed based on identified protein's molecular weight or isoelectric point between the two digestion methods. Implementation of the IMER into the proteomic workflow provided similar protein identification results, automation for sample analysis, and reduced the analysis time by 15h.
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
N-p-Tosyl-L-phenylalanine chloromethyl ketone, ≥97% (TLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein