MilliporeSigma
  • Circulation Enrichment of Functional Endothelial Progenitor Cells by Infantile Phototherapy.
Circulation Enrichment of Functional Endothelial Progenitor Cells by Infantile Phototherapy.
Journal of cellular biochemistry (2016-06-30)
Vahid Siavashi, Simin Asadian, Azam Sharifi, Masoumeh Esmaeilivand, Roohangiz Norouzinia, Mohammad Azadbakht, Seyed Mahdi Nassiri
ABSTRACT
Phototherapy is the most common therapy used for severe jaundice. There is increasing evidence that phototherapy can directly affect the expression and function of cell surface receptors including adhesion molecules, cytokines, and growth factor receptors. In this study, the effect of two infantile phototherapy regimens, including single and intensive phototherapy was investigated on biological features of circulation endothelial progenitor cells (cEPCs), as well as on serum secretion of two important chemotactic cytokines, SDF-1 and VEGF. Sixty infants diagnosed with severe hyperbilirubinemia and exposed to phototherapy were enrolled in this study. cEPCs were isolated before and after phototherapy and then migratory, proliferative, tubulogenic, and functional properties of these cells were analyzed. Our results revealed that intensive phototherapy markedly increased the release of EPCs into the circulation, and augmented the serum concentrations of both SDF-1 and VEGF cytokines. Cell proliferation, tubulogenic, and migratory properties of cEPCs isolated and expanded from infants with intensive phototherapy were significantly improved. cEPCs from infants with intensive phototherapy also showed greater levels of acetylated low-density lipoprotein and lectin binding. Overall, our results showed that the intensive phototherapy regimen can mobilize functional EPCs into the circulation through up-regulation of serum levels of VEGF and SDF-1, indicating phototherapy as an effective modality for improvement of stem cell mobilization in the therapeutic regenerative medicine. J. Cell. Biochem. 118: 330-340, 2017. © 2016 Wiley Periodicals, Inc.
Sigma-Aldrich
Ficoll® solution, Type 400, 20% in H2O
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), FITC conjugate, lyophilized powder
Sigma-Aldrich
DAPI, for nucleic acid staining