MilliporeSigma
  • Rheological characterization of solutions and thin films made from amylose-hexadecylammonium chloride inclusion complexes and polyvinyl alcohol.
Rheological characterization of solutions and thin films made from amylose-hexadecylammonium chloride inclusion complexes and polyvinyl alcohol.
Carbohydrate polymers (2017-02-13)
William T Hay, Jeffrey A Byars, George F Fanta, Gordon W Selling
ABSTRACT
The rheological properties of aqueous solutions and films made from blends of polyvinyl alcohol (PVOH) and amylose-hexadecylammonium chloride inclusion complexes (Hex-Am) were investigated to better understand the polymer interactions and processing parameters. Aqueous solutions of Hex-Am displayed non-Newtonian shear thinning characteristics, becoming highly viscous at 4.2% solids and forming a strong mechanical gel at 10% solids. Cationic Hex-Am was observed to have dramatically different rheological temperature response profiles from anionic amylose-sodium palmitate inclusion complexes, displaying a precipitous increase in viscosity upon cooling from 95°C to 50°C. Aqueous solution blends of 1:1 PVOH/Hex-Am lack this precipitous increase in viscosity, indicating that PVOH reduces amylose-chain entanglement. Films cast from varying blends of Hex-Am and PVOH were thermostable to 200°C, and displayed decreasing storage modulus with increasing concentrations of PVOH in film blends. Films cast from Hex-Am/PVOH absorb water vapor at lower rates than their constitutive polymers.
Sigma-Aldrich
Hexadecylamine, 98%
Sigma-Aldrich
Sodium palmitate, ≥98.5%