MilliporeSigma
  • Pre-cultivation with Selected Prebiotics Enhances the Survival and the Stress Response of
Pre-cultivation with Selected Prebiotics Enhances the Survival and the Stress Response of
Frontiers in microbiology (2017-07-01)
Mariantonietta Succi, Patrizio Tremonte, Gianfranco Pannella, Luca Tipaldi, Autilia Cozzolino, Rossana Romaniello, Elena Sorrentino, Raffaele Coppola
ABSTRACT
In our study, we dwelled upon combinations of lactobacilli/prebiotics, considering four different strains belonging to the
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Millipore
Bile bovine, dried, unfractionated
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mL
In Stock
Details...
$64.30
4 x 25 mL
In Stock
Details...
$83.90
1 L
In Stock
Details...
$146.00
2.5 L
In Stock
Details...
$223.00