MilliporeSigma
Endocytosis following dopamine D
Molecular psychiatry (2016-12-07)
N Shioda, Y Yabuki, Y Wang, M Uchigashima, T Hikida, T Sasaoka, H Mori, M Watanabe, M Sasahara, K Fukunaga
ABSTRACT
Aberrant dopamine D
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
MISSION® esiRNA, targeting human PDGFRB
Sigma-Aldrich
MISSION® esiRNA, targeting human DRD2