MilliporeSigma
  • Multiparametric approach with diffusion-weighted imaging and dynamic contrast-enhanced MRI: a comparison study for differentiating between benign and malignant bone lesions in adults.
Multiparametric approach with diffusion-weighted imaging and dynamic contrast-enhanced MRI: a comparison study for differentiating between benign and malignant bone lesions in adults.
Clinical radiology (2017-03-23)
E Oh, Y C Yoon, J H Kim, K Kim
ABSTRACT
To evaluate and compare the diagnostic performance of quantitative parameters derived from diffusion-weighted imaging (DWI) and dynamic contrast-enhanced magnetic resonance imaging (DCE-MRI) in differentiating benign and malignant bone tumours. Fifty-five patients (age range, 21-82 years; mean age, 55 years) underwent pretreatment MRI. Apparent diffusion coefficient (ADC) values were calculated by DWI. The DCE-MRI data were analysed for the volume transfer constant (K All parameter values (except V The K
Sigma-Aldrich
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, ≥97.0% (CHN)