Merck
  • Curvature of synthetic and natural surfaces is an important target feature in classical pathway complement activation.
Curvature of synthetic and natural surfaces is an important target feature in classical pathway complement activation.
Journal of immunology (Baltimore, Md. : 1950) (2010-01-08)
Martin Bjerregård Pedersen, Xingfei Zhou, Esben Kjaer Unmack Larsen, Uffe Skov Sørensen, Jørgen Kjems, Jens Vinge Nygaard, Jens Randel Nyengaard, Rikke Louise Meyer, Thomas Boesen, Thomas Vorup-Jensen
ABSTRACT
The binding of Abs to microbial surfaces followed by complement activation constitutes an important line of defense against infections. In this study, we have investigated the relationship between complement activation and the binding of human IgM Abs to surfaces with different curvatures. IgM Abs to dextran were shown to activate complement potently on dextran-coated particles having a diameter around 250 nm, whereas larger (600 nm) particles were less potent activators. This selectivity regarding particle dimension was also found for complement activation by colloidal substances of microbial origin. Peptidoglycan (PGN) is the major chemical component in the cell wall of Gram-positive bacteria. Fragments of purified PGN with sizes of approximately 100 nm promoted complement activation effectively through the classical pathway. By contrast, larger or smaller fragments of PGN did not activate complement strongly. A careful analysis of PGN fragments released during planctonic growth of Staphylococcus aureus showed that these include curvatures that would permit strong IgM-mediated complement activation, whereas the curvature of intact cells would be less effective for such activation. Consistently, we found that the suspended PGN fragments were strong activators of complement through the classical pathway. We suggest that these fragments act as decoy targets for complement activation, providing protection for S. aureus against the host immune response to infection.
Sigma-Aldrich
Zymosan A from Saccharomyces cerevisiae, for inducing inflamatory response
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mutanolysin from Streptomyces globisporus ATCC 21553, lyophilized powder, ≥4000 units/mg protein (biuret), Chromatographically purified
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mutanolysin from Streptomyces globisporus ATCC 21553, free of DNA contaminants, suitable for Microbiome research, lyophilized powder, ≥4000 units/mg protein (biuret)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein