MilliporeSigma
  • RYBP Expression Is Regulated by KLF4 and Sp1 and Is Related to Hepatocellular Carcinoma Prognosis.
RYBP Expression Is Regulated by KLF4 and Sp1 and Is Related to Hepatocellular Carcinoma Prognosis.
The Journal of biological chemistry (2016-12-29)
Qiaojiajie Zhao, Weihua Cai, Xuan Zhang, Shuo Tian, Junwen Zhang, Haibo Li, Congcong Hou, Xiaoli Ma, Hong Chen, Bingren Huang, Deng Chen
ABSTRACT
The expression of Ring1- and YY1-binding protein (RYBP) is reduced in several human cancers, but the molecular mechanism(s) have remained elusive. In this study, we used human hepatocellular carcinoma (HCC) cell lines and tissue specimens to study the mechanism and herein report several new findings. First, we cloned and characterized the basal promoter region of the human RYBP gene. We found that the decreased RYBP expression in HCC tissues was not due to promoter sequence variation/polymorphisms or CpG dinucleotide methylation. We identified two transcription factors, KLF4 and Sp1, which directly bind the promoter region of RYBP to induce and suppress RYBP transcription, respectively. We mapped the binding sites of KLF4 and Sp1 on the RYBP promoter. Studies in vitro showed that KLF4 suppresses whereas Sp1 promotes HCC cell growth through modulating RYBP expression. Deregulated KLF4 and Sp1 contributed to decreased expression of RYBP in HCC tumor tissues. Our studies of human HCC tissues indicated that a diminished RYBP level in the tumor (in association with altered KLF4 and Sp1 expression) was statistically associated with a larger tumor size, poorer differentiation, and an increased susceptibility to distant metastasis. These findings help to clarify why RYBP is decreased in HCC and indicate that deregulated KLF4, Sp1, and RYBP may lead to a poorer prognosis. Our findings support the idea that RYBP may represent a target for cancer therapy and suggest that it may be useful as a prognostic biomarker for HCC, either alone or in combination with KLF4 and Sp1.
Sigma-Aldrich
Monoclonal Anti-SP1 antibody produced in mouse, clone 4C8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-DEDAF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)