MilliporeSigma
  • CYP2E1 regulates the development of radiation-induced pulmonary fibrosis via ER stress- and ROS-dependent mechanisms.
CYP2E1 regulates the development of radiation-induced pulmonary fibrosis via ER stress- and ROS-dependent mechanisms.
American journal of physiology. Lung cellular and molecular physiology (2017-08-12)
Beomseok Son, TaeWoo Kwon, Sungmin Lee, IkJoon Han, Wanyeon Kim, HyeSook Youn, BuHyun Youn
ABSTRACT
Radiation-induced pulmonary fibrosis (RIPF) is one of the most common side effects of lung cancer radiotherapy. This study was conducted to identify the molecular mechanism responsible for RIPF. We revealed that the transcriptional level of cytochrome P450 2E1 (CYP2E1) was elevated by examining expression profile analysis of RIPF mouse models. We also confirmed that CYP2E1 regulated levels of endoplasmic reticulum (ER) stress and reactive oxygen species (ROS) in alveolar epithelial type II (AE2) cells and lung fibroblasts. Inhibition of CYP2E1 via its siRNA or inhibitor significantly attenuated epithelial-to-mesenchymal transition and apoptosis of AE2 cells, as well as myofibroblast formation induced by radiation. Finally, the effects of a CYP2E1 inhibitor on development of RIPF were evaluated by in vivo studies. Taken together, the results of the present study suggest that CYP2E1 is an important mediator of RIPF development that functions by increasing cellular ER stress and ROS levels.
Sigma-Aldrich
MISSION® esiRNA, targeting human CYP2E1
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00
Sigma-Aldrich
4-Phenylbutyric acid, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00