MilliporeSigma
  • Dynamic covalent bonding-triggered supramolecular gelation derived from tetrahydroxy-bisurea derivatives.
Dynamic covalent bonding-triggered supramolecular gelation derived from tetrahydroxy-bisurea derivatives.
Soft matter (2017-11-08)
Suansuan Wang, Kaiqiang Liu, Sheng Gao, Jinqiang Wang, Ravi Kumar Marella, Yu Fang
ABSTRACT
A new class of bisurea derivatives bearing tetrahydroxy groups have been proven to be non-gelators in water and various organic solvents even under long-term sonication or efficient heating treatment. We found that it is possible to trigger physical gelation behaviour by constructing dynamic covalent bonding. The results show that formation of dynamic covalent bonding between the borate anion and ethanediol substituent in these bisurea derivatives brings about rapid physical gelation at ambient temperature in a mixture of DMSO and water. During dynamic covalent bonding-triggered gelation, the stepgrowth polymerization from the B-O bonds would increase the size of the molecules and reduce the entropy of mixing as well as facilitate ion-dipole interactions in the linear polymeric gelators. They would drive a self-assembly transition and boost the construction of gel networks in coordination with α-tape urea-urea hydrogen bonding. The gelation mechanism was explored by
Sigma-Aldrich
1,4-Phenylene diisocyanate
Sigma-Aldrich
Serinol, 98%
Sigma-Aldrich
m-Xylylene diisocyanate, technical, ≥90% (GC)