MilliporeSigma
  • Effect of decaglycerol mono-oleate on chemiluminescence of human neutrophils.
Effect of decaglycerol mono-oleate on chemiluminescence of human neutrophils.
Luminescence : the journal of biological and chemical luminescence (1999-12-22)
Q Liu, K Suzuki, S Kudo, M Yamada, K Kowatari, T Umeda, S Nakaji, K Sugawara
ABSTRACT
Decaglycerol mono-oleate (DGMO), a type of food additive (a polyglycerol ester of fatty acids), was evaluated for its in vitro effect on reactive oxygen species (ROS) generation by human neutrophils using a luminol-dependent chemiluminescence assay. Neutrophils were isolated by the Histopaque-1077/1119 double density gradient technique. Opsonized zymosan (OZ) and phorbol 12-myristate 13-acetate (PMA) were employed as artificial stimuli. DGMO inhibited the chemiluminescence response of OZ-stimulated neutrophils by 58% (p < 0.01) at a concentration of 0.5 mg/mL. DGMO inhibited the PMA-induced chemiluminescence by 40% (p < 0.01) at a concentration of 0.01 mg/mL. The inhibition phenomena could not be attributed to light absorption by DGMO alone, because the percentage chemiluminescence in the presence of DGMO is lower than the percentage transmittance of DGMO at 425 nm. These results suggest that DGMO downregulates ROS generation by human neutrophils.
Sigma-Aldrich
Histopaque®-1077 Hybri-Max, liquid, sterile-filtered, BioReagent, suitable for hybridoma
Pricing and Inventory is not available. Please check the Millipore website for more information.