MilliporeSigma
  • Exercise and Doxorubicin Treatment Modulate Cardiac Mitochondrial Quality Control Signaling.
Exercise and Doxorubicin Treatment Modulate Cardiac Mitochondrial Quality Control Signaling.
Cardiovascular toxicology (2017-05-26)
I Marques-Aleixo, E Santos-Alves, J R Torrella, P J Oliveira, J Magalhães, A Ascensão
ABSTRACT
The cross-tolerance effect of exercise against heart mitochondrial-mediated quality control, remodeling and death-related mechanisms associated with sub-chronic Doxorubicin (DOX) treatment is yet unknown. We therefore analyzed the effects of two distinct chronic exercise models (endurance treadmill training-TM and voluntary free wheel activity-FW) performed during the course of the sub-chronic DOX treatment on mitochondrial susceptibility to permeability transition pore (mPTP), apoptotic and autophagic signaling and mitochondrial dynamics. Male Sprague-Dawley rats were divided into six groups (n = 6 per group): saline sedentary (SAL + SED), SAL + TM (12-weeks treadmill), SAL + FW (12-weeks voluntary free-wheel), DOX + SED [7-weeks sub-chronic DOX treatment (2 mg kg
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Available to ship on April 09, 2025
Details...
$116.00
100 g
Available to ship on April 09, 2025
Details...
$345.00
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Available to ship on April 09, 2025
Details...
$116.00
100 g
Available to ship on April 09, 2025
Details...
$345.00