MilliporeSigma
  • An intrinsic vasopressin system in the olfactory bulb is involved in social recognition.
An intrinsic vasopressin system in the olfactory bulb is involved in social recognition.
Nature (2010-02-26)
Vicky A Tobin, Hirofumi Hashimoto, Douglas W Wacker, Yuki Takayanagi, Kristina Langnaese, Celine Caquineau, Julia Noack, Rainer Landgraf, Tatsushi Onaka, Gareth Leng, Simone L Meddle, Mario Engelmann, Mike Ludwig
ABSTRACT
Many peptides, when released as chemical messengers within the brain, have powerful influences on complex behaviours. Most strikingly, vasopressin and oxytocin, once thought of as circulating hormones whose actions were confined to peripheral organs, are now known to be released in the brain, where they have fundamentally important roles in social behaviours. In humans, disruptions of these peptide systems have been linked to several neurobehavioural disorders, including Prader-Willi syndrome, affective disorders and obsessive-compulsive disorder, and polymorphisms of V1a vasopressin receptor have been linked to autism. Here we report that the rat olfactory bulb contains a large population of interneurons which express vasopressin, that blocking the actions of vasopressin in the olfactory bulb impairs the social recognition abilities of rats and that vasopressin agonists and antagonists can modulate the processing of information by olfactory bulb neurons. The findings indicate that social information is processed in part by a vasopressin system intrinsic to the olfactory system.
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
80 units
Estimated to ship on March 23, 2025
$364.00