MilliporeSigma
  • Trace analysis of ethanol, MTBE, and related oxygenate compounds in water using solid-phase microextraction and gas chromatography/mass spectrometry.
Trace analysis of ethanol, MTBE, and related oxygenate compounds in water using solid-phase microextraction and gas chromatography/mass spectrometry.
Analytical chemistry (2000-10-12)
D A Cassada, Y Zhang, D D Snow, R F Spalding
ABSTRACT
Solid-phase microextraction (SPME) and gas chromatography/mass spectrometry have been combined for trace-level determination of very polar compounds in water, including the widely used gasoline oxygenates ethanol and methyl tert-butyl ether (MTBE). A relatively simple extraction method using a divinylbenzene/Carboxen/poly(dimethylsiloxane) SPME fiber was optimized for the routine analysis of ethanol and MTBE in groundwater and reagent water. A sodium chloride concentration of 25% (w/w) combined with an extraction time of 25 min provided the greatest sensitivity while maintaining analytical efficiency. Replicate analyses in fortified reagent and groundwater spiked with microgram per liter concentrations of ethanol and MTBE indicate quantitative and reproducible recovery of these and related oxygenate compounds. Method detection limits were 15 microg L(-1) for ethanol, 1.8 microg L(-1) for tert-butyl alcohol, 0.038 microg L(-1) for tert-amyl methyl ether, 0.025 microg L(-1) for ethyl-tert-butyl ether, and 0.008 microg L(-1) for MTBE.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
ORBO-93 Sorbent Tube, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$260.00
$182.00