MilliporeSigma
  • The evolution of volatile compounds profile of "Toscano" dry-cured ham during ripening as revealed by SPME-GC-MS approach.
The evolution of volatile compounds profile of "Toscano" dry-cured ham during ripening as revealed by SPME-GC-MS approach.
Journal of mass spectrometry : JMS (2010-08-28)
C Pugliese, F Sirtori, L Calamai, O Franci
ABSTRACT
The volatile compounds profile is an important feature for the characterization of dry-cured hams. Some minor typical Italian products, such as 'Toscano' ham, have been poorly studied in regards to their composition of volatile compounds. In this article, we studied the evolution of the aromatic profile of 'Toscano' dry-cured ham by solid-phase microextraction-gas chromatographic-mass spectrometry (SPME-GC-MS) with ripening. Ten right thighs were cured according to the 'Toscano' PDO protocol, sampled at 0, 1, 3, 6 and 12 months and submitted to volatile compounds analysis by SPME with a Divinylbenzene (DVB)/Carboxen/Polydimethylsiloxane (PDMS) 75-µ Stable Flex fibre. An Agilent 5975C mass selective detector (MSD) spectrometer with electron ionization (EI) source operating in scan mode within the m/z 29-350 range was used for data collection. Seven internal standards, either deuterium labeled or absent in the specimens and chosen to represent low or high boiling esters, alcohols, acids or phenols, were added to the homogenized samples and used to normalize the SPME fibre response to account for response changes upon wearing. Linear calibrations were obtained in this way for selected representative compounds. Over 60 compounds belonging to esters, aldehydes, organic acids, ketones and alcohols were identified by comparison with spectral libraries and Kovats indices. Aldehydes were the most represented chemical family, followed by organic acids, alcohols, ketones and esters. The aldehydes and ketones increased during the first 3 months, when the larger formation of volatiles occurred. For other families, the evolution over time was less evident. The principal component and discriminant analyses of the aromatic profile were effective in classifying the hams at 0, 6 or 12 months of ripening while for 1 and 3 months' samples a partial overlapping was shown. These results represent the first characterization of 'Toscano' ham and may constitute the basis to identify the best ripening time and define an analytical quality standard for this typical ham.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
ORBO-93 Sorbent Tube, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$40.20
100 g
In Stock
Details...
$81.20