MilliporeSigma
GC Analysis of a 14-Component Polybrominated Diphenyl Ether (PBDE) Mix on SLB®-5ms
GC Analysis of a 14-Component Polybrominated Diphenyl Ether (PBDE) Mix on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
125 °C (1 min), 10 °C/min to 340 °C (15 min)
inj. temp.
300 °C
MSD interface
340 °C
scan range
SIM
carrier gas
helium, 1.5 mL/min, constant
injection
1 μL, splitless, pulsed (30 psi until 0.2 min)
liner
4 mm I.D. single taper
sample
PBDE standard, 2.5-10 μg/mL in n-nonane
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany