GC Analysis of Blood Alcohols in Human Plasma on SUPELCOWAX™ 10 after SPME using 60 μm Carbowax® Fiber
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
sample/matrix
blood alcohols at concentrations indicated in human plasma
SPME fiber
60 μm Carbowax
extraction
headspace, 50 °C (5 min)
desorption process
220 °C, 2 min
column
SUPELCOWAX 10, 30 m × 0.25 mm I.D., 0.50 μm (24284)
oven
35 °C (2 min), 10 °C/min to 125 °C (1 min)
detector
FID, 200 °C
carrier gas
helium, 1.0 mL/min constant
injection
10:1 split
liner
0.75 mm I.D. SPME liner
Description
Legal Information
Carbowax is a registered trademark of Union Carbide
SUPELCOWAX is a trademark of Sigma-Aldrich Co. LLC