Merck
GC Analysis of Impurities in MTBE on Petrocol® DH
GC Analysis of Impurities in MTBE on Petrocol® DH suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Petrocol® DH Capillary GC Column
L × I.D. 100 m × 0.25 mm, df 0.50 μm
CONDITIONS
column
Petrocol DH, 100 m × 0.25 mm I.D., 0.50 μm (24160-U)
oven
50 °C (13 min), 10 °C/min to 180 °C
inj. temp.
250 °C
detector
FID, 310 °C
carrier gas
helium, 20 cm/sec @ 35 °C
injection
1 μL, 200:1 split
sample
MTBE Contaminants Mix A (47942)
Description
Legal Information
Petrocol is a registered trademark of Merck KGaA, Darmstadt, Germany