MilliporeSigma
GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms
GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
45 °C (3 min), 20 °C/min to 360 °C (10 min)
inj. temp.
275 °C
detector
FID, 365 °C
carrier gas
helium, 1.3 mL/min. constant
injection
1.0 μL, 100:1 split
liner
2 mm I.D. straight
sample
TPH Mix 3, each analyte at 1000 μg/mL in carbon disulfide (861394-U)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany