MilliporeSigma
GC Analysis of Volatiles in PVC Pipe on SPB®-HAP after Collection/Desorption using Carbotrap® 300 Tube
GC Analysis of Volatiles in PVC Pipe on SPB®-HAP after Collection/Desorption using Carbotrap® 300 Tube suitable for GC, application for air monitoring
Materials
related product
Product No.
Description
Pricing
ATIS Adsorbent Tube Injector System 110V
ATIS Adsorbent Tube Injector System 230V
ATIS Extraction Glassware with Micro Connector
CONDITIONS
sample/matrix
(1g PVC pipe Schedule 40)
thermal desorption tube
Carbotrap® 300 Glass-Fritted Thermal Desorption Tube, 1/4 in O.D. x 3.5 in L (30141-U)
dry purge
60 min
desorption process
350 °C with 10 min hold
transfer line/valve temp.
180 °C
column
SPB®-HAP, 60 m x 0.32 mm I.D., 4 μm (25020-U)
oven
40 °C (3 min), 12 °C/min to 200 °C (20 min)
detector
MSD
MSD interface
210 °C
carrier gas
helium
Description
Analysis Note
Qualitative volatile headspace outgassing of PVC pipe using the ATIS with analysis by thermal desorption.
Legal Information
Carbotrap is a registered trademark of Merck KGaA, Darmstadt, Germany, SPB is a registered trademark of Merck KGaA, Darmstadt, Germany